Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate Dihydrate (Powder/USP/FCC), Fisher Chemical

Click to view available options
Quantity:
3kg
Packaging:
Poly Pail
6132-04-3
C6H9Na3O9
294.10
MFCD00150031
NLJMYIDDQXHKNR-UHFFFAOYSA-K
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
71474
CHEBI:32142
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
6132-04-3
C6H9Na3O9
294.10
MFCD00150031
NLJMYIDDQXHKNR-UHFFFAOYSA-K
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
71474
CHEBI:32142
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
Density 0.75 G/cm × 2 min.
Melting Point 300°C
Residual Solvents Meets Requirements (USP)
Water 10.0 to 13.0% (FCC/USP)
Color White
pH 7.5 to 9.5
Identification Pass Test (FCC/USP)
Packaging Poly Pail
Quantity 3kg
Alkalinity Pass Test (FCC/USP)
Decomposition Information >230°C
Linear Formula NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O
Formula Weight 294.1
Physical Form Solid
Percent Purity 99.0 to 100.5% (USP)
Grade FCC/USP
Chemical Name or Material Sodium Citrate Dihydrate
Show More Show Less

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0