Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate Dihydrate, Purified Trisodium Citrate Dihydrate, Reagents

Catalog No. p-200020884
Click to view available options
Quantity:
100 g
500 g

Sodium Citrate Dihydrate, Purified Trisodium Citrate Dihydrate, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6132-04-3
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym Trisodium Citrate Dihydrate, 2-Hydroxy-1,2,3-propanetricarboxylic Acid Trisodium Salt Dihydrate
IUPAC Name trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
SMILES O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Melting Point >300°C
Color Colorless/White
pH 7.5 to 9.0
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 1.665
Chemical Name or Material Sodium Citrate Dihydrate, Purified Trisodium Citrate Dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.