Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Cobaltinitrite, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
13600-98-1
Na3Co(NO2)6
13600-98-1
Na3Co(NO2)6

High quality chemicals for laboratory and specialized industrial use

    For Determination of Potassium
Formula Weight 403.94
Physical Form Powder
Packaging Poly Bottle
Quantity 125g
Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Sodium Cobaltinitrite