Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium cyanide, 98+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
5g
1kg
Packaging:
Glass bottle
143-33-9
CNNa
49
MFCD00003523
MNWBNISUBARLIT-UHFFFAOYSA-N
sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
8929
sodium;cyanide
[C-]#N.[Na+]
143-33-9
CNNa
49
MFCD00003523
MNWBNISUBARLIT-UHFFFAOYSA-N
sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
8929
sodium;cyanide
[C-]#N.[Na+]
C-X bond formation reagent
Assay Percent Range 98% min. (Argentometry)
Linear Formula NaCN
Beilstein 02, IV, 50
Fieser 04,446; 05,506; 06,535; 07,333; 08,453; 11,481; 13,185
Merck Index 15, 8741
Solubility Information Solubility in water: 370g/L (20°C). Other solubilities: sparingly soluble in ethanol
Formula Weight 49
Physical Form Crystals
Percent Purity 98+%
Grade Extra Pure
Cyanide cyanide: 52.0% to 52.9%
Packaging Glass bottle
Sodium Carbonate 0.2 to 0.8%
Sodium Hydroxide sodium hydroxide: 0.30% to 0.80%
Sodium Formate sodium formate: 0.10% to 0.50%
Solubility Solubility in water: 370g/L (20°C). Other solubilities: sparingly soluble in ethanol
Water 0.1% max.
Boiling Point 1497.0°C
Color White
Melting Point 562.0°C
Quantity 5g
Chemical Name or Material Sodium cyanide, 98+%
Show More Show Less

GHS H Statement
May be corrosive to metals.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lastin

GHS P Statement
Absorb spillage to prevent material damage.
Wear protective gloves/protective clothing.
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a p

GHS Signal Word: Danger

EINECSNumber : 205-599-4

Recommended Storage : Poison Room