Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Cyanide (Granular/Certified ACS), Fisher Chemical™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Specifications
Boiling Point | 1497°C |
Melting Point | 562°C |
Color | White |
pH | 11 to 12 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 49.01 |
Percent Purity | ≥95% |
Grade | Certified ACS |
Identification | Pass Test |
Show More |
Safety and Handling
DOTInformation | DOT Class 6.1, : Poison |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction