Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium cyanide, 98%, for analysis, Acros Organics

Click to view available options
Quantity:
500g
1kg
Packaging:
Plastic bottle
143-33-9
CNNa
49.008
MNWBNISUBARLIT-UHFFFAOYSA-N
sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
8929
sodium;cyanide
[C-]#N.[Na+]
143-33-9
CNNa
49.008
MNWBNISUBARLIT-UHFFFAOYSA-N
sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
8929
sodium;cyanide
[C-]#N.[Na+]
Assay Percent Range 98%
Linear Formula NaCN
Fieser 04,446; 05,506; 06,535; 07,333; 08,453; 11,481; 13,185
Formula Weight 49
Physical Form Granular Crystalline Powder
Percent Purity ≥97%
Grade Analysis
Chloride 0.025% max.
Copper (Cu) 0.002% max.
Insoluble in Water 0.01% max.
Iron (Fe) 0.01% max.
Lead (Pb) 0.001% max.
Packaging Plastic bottle
Phosphate 0.01% max.
Potassium (K) 0.2% max.
Sulfate 0.025% max.
Sulfide 0.003% max.
Thiocyanate 0.05% max.
Zinc (Zn) 0.02% max.
Color White
Quantity 1kg
Chemical Name or Material Sodium cyanide, p.a.
Show More Show Less

GHS H Statement
May be corrosive to metals. Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. Contact with acids liberates very toxic gas.

GHS P Statement
Absorb spillage to prevent material damage. Wear protective gloves/protective clothing. Rinse mouth. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician.

Danger

EINECSNumber : 205-599-4

RTECSNumber : VZ7525000

TSCA : TSCA