Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium cyclamate, 99%, Acros Organics

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Plastic bottle
139-05-9
C6H12NNaO3S
201.22
MFCD00003827
UDIPTWFVPPPURJ-UHFFFAOYSA-M
sodium cyclamate,sodium cyclohexylsulfamate,sodium n-cyclohexylsulfamate,cyclamate sodium,cyclamic acid sodium salt,assugrin,ibiosuc,suessette,suestamin,sugarin
23665706
CHEBI:82431
sodium;N-cyclohexylsulfamate
C1CCC(CC1)NS(=O)(=O)[O-].[Na+]
139-05-9
C6H12NNaO3S
201.22
MFCD00003827
UDIPTWFVPPPURJ-UHFFFAOYSA-M
sodium cyclamate,sodium cyclohexylsulfamate,sodium n-cyclohexylsulfamate,cyclamate sodium,cyclamic acid sodium salt,assugrin,ibiosuc,suessette,suestamin,sugarin
23665706
CHEBI:82431
sodium;N-cyclohexylsulfamate
C1CCC(CC1)NS(=O)(=O)[O-].[Na+]
Flash Point >200°C
Quantity 250g
Color White
Packaging Plastic bottle
Loss on Drying 1% max.
Infrared Spectrum Authentic
pH 5.5 to 7.5 (10% soln.)
Assay Percent Range 99%
Linear Formula C6H12NNaO3S
Solubility Information Solubility in water: 200g/L (20°C). Other solubilities: insoluble in ether and benzene
Formula Weight 201.22
Physical Form Powder
Percent Purity ≥98.5% (on dry substance)
Chemical Name or Material Sodium cyclamate
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.