Alfa Aesar™ Sodium cyclohexanebutyrate, AAS, Certified Na Content

Manufacturer: Alfa Aesar™ 8726902

Quantity 0.5g

 View more versions of this product

Catalog No. AA8726902

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 61886-29-1
    Molecular Formula C10H17NaO2
    Formula Weight 192.23
    MDL Number MFCD00036395
    Synonym Cyclohexanebutyric acid sodium salt; Sodium 4-cyclohexylbutyrate
    Chemical Name or Material Sodium cyclohexanebutyrate, Certified Na Content
    Grade AAS Grade

    Specifications

    Formula Weight 192.23
    Molecular Formula C10H17NaO2
    MDL Number MFCD00036395
    CAS 61886-29-1
    Synonym Cyclohexanebutyric acid sodium salt; Sodium 4-cyclohexylbutyrate
    Melting Point 264°C (dec.)
    TSCA TSCA
    Physical Form Powder
    Quantity 0.5g
    Chemical Name or Material Sodium cyclohexanebutyrate, Certified Na Content
    Grade AAS Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit