Sodium deoxycholate monohydrate, 98%, Alfa Aesar™

Manufacturer: Alfa Aesar B2075914

Quantity 25g

 View more versions of this product

Catalog No. AAB2075914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 145224-92-6
    Physical Form Powder
    Assay Percent Range 0.98
    Molecular Formula C24H39NaO.H2O
    Formula Weight 432.58; 414.58 (anhydrous)
    Chemical Name or Material Sodium Deoxycholate Monohydrate

    Specifications

    Formula Weight 432.58; 414.58 (anhydrous)
    Melting Point >300°C
    MDL Number MFCD00150750
    Synonym Deoxycholic acid sodium salt monohydrate
    Quantity 25g
    Molecular Formula C24H39NaO.H2O
    Physical Form Powder
    Assay Percent Range 0.98
    CAS 145224-92-6
    Chemical Name or Material Sodium Deoxycholate Monohydrate

    Description

    Applications
    Sodium Deoxycholate Monohydrate is used as a solubilization agent for estrogen synthetase from human placental microsomes. It is also used to study the immobilization of lipoxygenase in an alginate-silicate gel matrix and to investigate the in vitro dissolution of gall stones.

    Solubility
    Soluble in water.

    Notes
    Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit