Alfa Aesar™ Sodium deuteroxide, 30% w/w solution in D2O, 99.5% (Isotopic)

NMR Solvent

Manufacturer: Alfa Aesar™ 4234518

Quantity 50g

 View more versions of this product

Catalog No. AA4234518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14014-06-3
    Formula Weight 41.01
    MDL Number MFCD00037669
    UN Number UN1824
    Chemical Name or Material Sodium deuteroxide, Isotopic

    Specifications

    Formula Weight 41.01
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 1.462g/mL
    MDL Number MFCD00037669
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Sensitivity Air & Moisture Sensitive
    Refractive Index 1.4039
    UN Number UN1824
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE SOLUTION
    Physical Form Liquid
    Quantity 50g
    Concentration 30% w/w solution in D2O
    Health Hazard 1 Danger
    Odor Odorless
    CAS 14014-06-3
    Chemical Name or Material Sodium deuteroxide, Isotopic
    Assay 99.5%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit