Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium deuteroxide, for NMR, 40 wt.% solution in D2O, 99+% atom D, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Plastic bottle
14014-06-3,7789-20-0
HNaO
41.003
MFCD00037669
HEMHJVSKTPXQMS-DYCDLGHISA-M
sodium deuteroxide,sodium hydroxide na od,sodium deuteroxide, 40 wt% in deuterium oxide,naod,sodium hydroxide-d,sodium deuteroxide solution,sodium deuteroxide, 1 m in deuterium oxide,sodium deuteroxide, 0.1 m in deuterium oxide,sodium deuteroxide, 30 wt% in deuterium oxide,sodium hydroxide na od 6ci,7ci,8ci,9ci
23676750
[OH-].[Na+]
14014-06-3,7789-20-0
HNaO
41.003
MFCD00037669
HEMHJVSKTPXQMS-DYCDLGHISA-M
sodium deuteroxide,sodium hydroxide na od,sodium deuteroxide, 40 wt% in deuterium oxide,naod,sodium hydroxide-d,sodium deuteroxide solution,sodium deuteroxide, 1 m in deuterium oxide,sodium deuteroxide, 0.1 m in deuterium oxide,sodium deuteroxide, 30 wt% in deuterium oxide,sodium hydroxide na od 6ci,7ci,8ci,9ci
23676750
[OH-].[Na+]
Color Colorless
Density 1.4600g/mL
Deuterium (D) 99.0% min.
Quantity 50g
Packaging Plastic bottle
Linear Formula NaOD
Refractive Index 1.4039
Formula Weight 41
Physical Form Liquid
Grade NMR
Chemical Name or Material Sodium deuteroxide, 99+ atom % D
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 237-825-2