Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium dichloroacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
2156-56-1
C2HCl2NaO2
150.92
MFCD00070489
LUPNKHXLFSSUGS-UHFFFAOYSA-M
sodium dichloroacetate,sodium 2,2-dichloroacetate,dichloroacetic acid sodium salt,acetic acid, dichloro-, sodium salt,ceresine,dichloroacetate sodium,dichloroctan sodny czech,sodium dichloroacetate usan,ccris 7697,dichloroacetic acid, sodium salt
517326
sodium;2,2-dichloroacetate
C(C(=O)[O-])(Cl)Cl.[Na+]
2156-56-1
C2HCl2NaO2
150.92
MFCD00070489
LUPNKHXLFSSUGS-UHFFFAOYSA-M
sodium dichloroacetate,sodium 2,2-dichloroacetate,dichloroacetic acid sodium salt,acetic acid, dichloro-, sodium salt,ceresine,dichloroacetate sodium,dichloroctan sodny czech,sodium dichloroacetate usan,ccris 7697,dichloroacetic acid, sodium salt
517326
sodium;2,2-dichloroacetate
C(C(=O)[O-])(Cl)Cl.[Na+]
Assay Percent Range 98%
Linear Formula NaOC(O)CH(Cl)2
Beilstein 02, III, 454
Solubility Information Solubility in water: soluble
Formula Weight 150.92
Physical Form Powder
Percent Purity ≥97.5%
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Melting Point 198°C
Quantity 50g
Chemical Name or Material Sodium dichloroacetate, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 218-461-3

RTECSNumber : AG9275000