Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dichromate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7789-12-0
Cr2H4Na2O9
297.995
JYDRNIYTFCBIFC-UHFFFAOYSA-N
108005

High quality chemicals for laboratory and specialized industrial use

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
SMILES O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
Molecular Weight (g/mol) 297.995
PubChem CID 108005
Formula Weight 298
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.35kg
Quantity 500g
Chemical Name or Material Sodium Dichromate, Dihydrate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less