Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dichromate Dihydrate, Purified, Reagents

Catalog No. 01337025
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
50 lb.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-025 50 lb.
01-337-028 10 kg
01-337-027 2.5 kg
01-337-026 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-025 Supplier Reagents Holdings Llc Supplier No. C231600050F5
Only null left
Add to Cart
Add to Cart

Sodium Dichromate Dihydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
Molecular Weight (g/mol) 298.00
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym Sodium Bichromate
IUPAC Name disodium [(oxidodioxochromio)oxy]chromiumoylolate dihydrate
SMILES O.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

Specifications

Boiling Point 400°C
Melting Point 357°C
Color Orange/Red
Physical Form Solid
Quantity 50 lb.
Grade Pure
Packaging Fiber Drum
Specific Gravity 2.35
Chemical Name or Material Sodium Dichromate Dihydrate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.