Sodium dichromate dihydrate, Reagent Grade, Alfa Aesar™

Manufacturer: Alfa Aesar 1261136

Quantity 500g

 View more versions of this product

Catalog No. AA1261136

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7789-12-0
    Physical Form Crystalline
    Molecular Formula Cr2Na2O7·2H2O
    Formula Weight 298.00 (261.97 Anhydrous)
    MDL Number MFCD00149166
    Synonym Sodium bichromate
    Chemical Name or Material Sodium dichromate dihydrate
    Grade Reagent Grade

    Specifications

    Formula Weight 298.00 (261.97 Anhydrous)
    Quantity 500g
    Color Red
    DOT Information DOT Class: 5.1, Packaging Group: II
    EINECS Number 234-190-3
    Physical Form Crystalline
    Molecular Formula Cr2Na2O7·2H2O
    MDL Number MFCD00149166
    CAS 7789-12-0
    Synonym Sodium bichromate
    Appearance Red crystals
    Health Hazard 1 H272-H301-H330-H312-H317-H334-H340-H350-H360-H372-H314-H400-H410: May intensify fire; oxidizer. Toxic if swallowed. Fatal if inhaled. Harmful in contact with skin. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Sodium dichromate dihydrate
    Grade Reagent Grade

    H272-H301-H330-H312-H317-H334-H340-H350-H360-H372-H314-H400-H410: May intensify fire; oxidizer. Toxic if swallowed. Fatal if inhaled. Harmful in contact with skin. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit