Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium dicyanamide, 97%, pure, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
1934-75-4
C2N3Na
89.03
MFCD00003535
IXBPPZBJIFNGJJ-UHFFFAOYSA-N
sodium dicyanamide, sodium dicyanoazanide, sodium dicyanoamide, unii-i9c96i1mre, cyanamide, cyano-, sodium salt, cyanamide,cyano-,sodium salt, i9c96i1mre, cyanamide, n-cyano-, sodium salt 1:1, dicyanamide, sodium salt, nadca
4350929
sodium;dicyanoazanide
C(#N)[N-]C#N.[Na+]
1934-75-4
C2N3Na
89.03
MFCD00003535
IXBPPZBJIFNGJJ-UHFFFAOYSA-N
sodium dicyanamide, sodium dicyanoazanide, sodium dicyanoamide, unii-i9c96i1mre, cyanamide, cyano-, sodium salt, cyanamide,cyano-,sodium salt, i9c96i1mre, cyanamide, n-cyano-, sodium salt 1:1, dicyanamide, sodium salt, nadca
4350929
sodium;dicyanoazanide
C(#N)[N-]C#N.[Na+]
Melting Point >300°C
Packaging Plastic bottle
Quantity 500g
Water 0.5% max.
Color Pink or White
Infrared Spectrum Authentic
Assay Percent Range 2% max. (NaCl) (Argentometry)
Linear Formula NaN(CN)2
Beilstein 03, III, 154
Solubility Information Solubility in water: 260g/L (30°C). Other solubilities: insoluble in aceton,etoh and ethyl acetate
Formula Weight 89.03
Physical Form Powder
Percent Purity 97%
Grade Pure
Chemical Name or Material Sodium dicyanamide, Pure
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lens

GHS Signal Word: Warning

EINECSNumber : 217-703-5