Alfa Aesar™ Sodium diethyldithiocarbamate trihydrate, ACS

Manufacturer: Alfa Aesar™ 3337922

Quantity 100g

 View more versions of this product

Catalog No. AA3337922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20624-25-3
    Formula Weight 225.31 (171.28 Anhydrous)
    MDL Number MFCD00150617
    Beilstein 3920507
    Merck Index 14,3378
    Synonym Diethyldithiocarbamic acid sodium salt trihydrate
    Chemical Name or Material Sodium diethyldithiocarbamate trihydrate
    Grade ACS Grade

    Specifications

    Formula Weight 225.31 (171.28 Anhydrous)
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    RTECS Number EZ6550000
    CAS 20624-25-3
    Beilstein 3920507
    Color White to off-white
    Quantity 100g
    Sensitivity Hygroscopic
    MDL Number MFCD00150617
    Merck Index 14,3378
    Melting Point 94° to 102°C
    Health Hazard 1 Warning
    TSCA TSCA
    Synonym Diethyldithiocarbamic acid sodium salt trihydrate
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Chemical Name or Material Sodium diethyldithiocarbamate trihydrate
    Grade ACS Grade

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit