Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Diethyldithiocarbamate, Trihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
20624-25-3
C5H16NNaO3S2
225.297
WWGXHTXOZKVJDN-UHFFFAOYSA-M
sodium diethyldithiocarbamate trihydrate,dithiocarb trihydrate,cupral,diethyldithiocarbamic acid sodium salt trihydrate,ditiocarb sodium trihydrate,unii-04574exf2c,diethyldithiocarbamate sodium trihydrate,carbamic acid, diethyldithio-, sodium salt, trihydrate,diethyldithiocarbamate sodium salt trihydrate,diethyldithiokarbaman sodny trihydrat czech
517546
sodium;N,N-diethylcarbamodithioate;trihydrate
CCN(CC)C(=S)[S-].O.O.O.[Na+]
20624-25-3
C5H16NNaO3S2
225.297
WWGXHTXOZKVJDN-UHFFFAOYSA-M
sodium diethyldithiocarbamate trihydrate,dithiocarb trihydrate,cupral,diethyldithiocarbamic acid sodium salt trihydrate,ditiocarb sodium trihydrate,unii-04574exf2c,diethyldithiocarbamate sodium trihydrate,carbamic acid, diethyldithio-, sodium salt, trihydrate,diethyldithiocarbamate sodium salt trihydrate,diethyldithiokarbaman sodny trihydrat czech
517546
sodium;N,N-diethylcarbamodithioate;trihydrate
CCN(CC)C(=S)[S-].O.O.O.[Na+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 225.31
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.10kg
Quantity 500g
Chemical Name or Material Sodium Diethyldithiocarbamate, Trihydrate