Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Diethyldithiocarbamate, Trihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle

Chemical Identifiers

20624-25-3
C5H16NNaO3S2
225.297
WWGXHTXOZKVJDN-UHFFFAOYSA-M
517546

High quality chemicals for laboratory and specialized industrial use

SMILES CCN(CC)C(=S)[S-].O.O.O.[Na+]
IUPAC Name sodium;N,N-diethylcarbamodithioate;trihydrate
Formula Weight 225.31
Packaging Poly Bottle
Specific Gravity 1L = 1.10kg
Quantity 125g
Chemical Name or Material Sodium Diethyldithiocarbamate, Trihydrate
Grade Baker Analyzed™ ACS Reagent