Alfa Aesar™ Sodium diphenylamine-4-sulfonate, 0.005M solution in water

Redox indicator

Manufacturer: Alfa Aesar™ 42809AP

Quantity 500mL

 View more versions of this product

Catalog No. AA42809AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6152-67-6
    Molecular Formula C12H10NNaO3S
    Formula Weight 271.27
    MDL Number MFCD00036468
    Chemical Name or Material Sodium diphenylamine-4-sulfonate

    Specifications

    Formula Weight 271.27
    Physical Form Liquid
    CAS 6152-67-6
    Molecular Formula C12H10NNaO3S
    Concentration 0.005M solution in water
    Quantity 500mL
    TSCA TSCA
    MDL Number MFCD00036468
    Odor Odorless
    Chemical Name or Material Sodium diphenylamine-4-sulfonate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit