Learn More
Sodium diphenylamine-4-sulfonate, ACS, Thermo Scientific Chemicals
Description
Redox indicatorSodium diphenylamine-4-sulfonate is used as a pH independent redox indicator used in the determination of redox potential. It is used in the preparation of poly-(aninile-co-N-(4-sulfophenyl)aniline) by polymerization with aniline. It acts as a ligand and form complexes with metal halides like lanthanum(III) chloride. It is also involved in the synthesis of gold nano crystals by reaction with chloroauric acid in the presence of polyethylene glycol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 2 g |
Solubility Information | Soluble in water. |
Formula Weight | 271.27 |
Grade | ACS |
Physical Form | Powder |
Chemical Name or Material | Sodium diphenylamine-4-sulfonate |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 228-165-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.