Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Diphenylaminesulfonate, ACS Reagent, Reagents
Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Color | Off-White |
Physical Form | Solid |
Quantity | 5 g |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 1 |
Chemical Name or Material | Sodium Diphenylaminesulfonate, ACS Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction