Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium DL-2-Hydroxybutyrate 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
5094-24-6
C4H7NaO3
126.09
MFCD00004565
MOSCXNXKSOHVSQ-UHFFFAOYNA-M
sodium 2-hydroxybutanoate, sodium 2-hydroxybutyrate, sodium dl-2-hydroxybutyrate, 2-hydroxybutyric acid sodium salt, dl-2-hydroxybutyric acid sodium salt, sodium hydroxybutyrate, acmc-1btqj, sodium, a-hydroxybutyrate, sodium 2-oxidanylbutanoate, sodiumdl-2-hydroxybutyrate
23663641
sodium 2-hydroxybutanoate
[Na+].CCC(O)C([O-])=O
5094-24-6
C4H7NaO3
126.09
MFCD00004565
MOSCXNXKSOHVSQ-UHFFFAOYNA-M
sodium 2-hydroxybutanoate, sodium 2-hydroxybutyrate, sodium dl-2-hydroxybutyrate, 2-hydroxybutyric acid sodium salt, dl-2-hydroxybutyric acid sodium salt, sodium hydroxybutyrate, acmc-1btqj, sodium, a-hydroxybutyrate, sodium 2-oxidanylbutanoate, sodiumdl-2-hydroxybutyrate
23663641
sodium 2-hydroxybutanoate
[Na+].CCC(O)C([O-])=O
Color White
Quantity 25g
Formula Weight 126.09
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material Sodium DL-2-Hydroxybutyrate