Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate, 20% (w/v), BiotechGrade, Spectrum™

Click to view available options
Quantity:
100 mL
200 mL
500 mL
Packaging:
Poly Bottle
151-21-3,7732-18-5
C12H25NaO4S
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
151-21-3,7732-18-5
C12H25NaO4S
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

S3008

Spectrum™ Sodium Dodecyl Sulfate, 20 Percent (w/v), BiotechGrade - Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

CAS Max % 0.2,0.8
Packaging Poly Bottle
Grade Biotech
pH 5.0 to 8.0
Quantity 200 mL