Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate, 95%, BAKER™, J.T. Baker™

Catalog No. p-5391660
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Quantity 500 g
Flash Point 170.00°C
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 0.40kg
Formula Weight 288.38
Percent Purity 95%
Chemical Name or Material Sodium Dodecyl Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.