Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Manufacturer:  J.T.Baker 409500

Packaging POLYSTORMOR
Quantity 25g

 View more versions of this product

Catalog No. 02-004-083

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

    For Electrophoresis and Protein Solubilization
Description & Specifications

Chemical Identifiers

151-21-3
288.38
Sodium Dodecyl Sulfate
CH3(CH2)11OSO3Na
SDS
Ultrapure Bioreagent

Specifications

151-21-3
288.38
SDS
25g
Sodium Dodecyl Sulfate
CH3(CH2)11OSO3Na
POLYSTORMOR
170.00°C
1L = 0.40kg
Ultrapure Bioreagent
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit