Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium fluoride, 97%, extra pure, ACROS Organics™

Click to view available options
Quantity:
25g
500g
1kg
Packaging:
Plastic bottle
7681-49-4
FNa
41.98
MFCD00003524
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium flouride, antibulit, floridine, fluonatril
5235
CHEBI:28741
sodium;fluoride
[F-].[Na+]
7681-49-4
FNa
41.98
MFCD00003524
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium flouride, antibulit, floridine, fluonatril
5235
CHEBI:28741
sodium;fluoride
[F-].[Na+]
C-X bond formation reagent
Assay Percent Range 97%
Linear Formula NaF
Fieser 01,1073; 02,382
Merck Index 15, 8754
Solubility Information Solubility in water: 40g/L (25°C). Other solubilities: slightly soluble in alcohol
Formula Weight 41.98
Physical Form Powder or Cyrstals
Percent Purity 97%
Grade Extra Pure
Chloride 120ppm max.
Heavy Metals (as Pb) 100ppm max.
Iron (Fe) 200ppm max.
Packaging Plastic bottle
Sodium Fluosilicate 1% max. (Na2SiF6)
Solubility Solubility in water: 40g/L (25°C). Other solubilities: slightly soluble in alcohol
Sulfate 5000ppm max.
Boiling Point 1700.0°C
Color White
Melting Point 993.0°C
Quantity 1kg
Chemical Name or Material Sodium fluoride, 97%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Contact with acids liberates very toxic gas.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs

GHS Signal Word: Danger

EINECSNumber : 231-667-8

RTECSNumber : WB0350000

TSCA : TSCA