Sodium fluoride, Optical Grade, Alfa Aesar™

Manufacturer: Alfa Aesar 1102309

Quantity 10g

 View more versions of this product

Catalog No. AA1102309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7681-49-4
    Physical Form Broken Crystals
    Molecular Formula NaF
    Formula Weight 41.99
    MDL Number MFCD00003524
    Chemical Name or Material Sodium fluoride
    Grade Optical Grade

    Specifications

    Physical Form Broken Crystals
    Formula Weight 41.99
    Quantity 10g
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: III
    Density 2.558g/cm3 at 20°C
    EINECS Number 231-667-8
    CAS 7681-49-4
    Odor Odorless
    Molecular Formula NaF
    Appearance White crystals
    Health Hazard 1 H300-H315-H319: Fatal if swallowed. Causes skin irritation. Causes serious eye irritation.
    MDL Number MFCD00003524
    Health Hazard 2 Do not breathe dust. Wear suitable protective clothing. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
    Chemical Name or Material Sodium fluoride
    Grade Optical Grade

    Do not breathe dust. Wear suitable protective clothing. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

    H300-H315-H319: Fatal if swallowed. Causes skin irritation. Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit