Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Fluoride, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

7681-49-4
FNa
41.988
PUZPDOWCWNUUKD-UHFFFAOYSA-M
5235
CHEBI:28741

High quality chemicals for laboratory and specialized industrial use

SMILES [F-].[Na+]
IUPAC Name sodium;fluoride
Formula Weight 41.99
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 2.78kg
Quantity 2.5kg
Chemical Name or Material Sodium Fluoride
Grade Baker Analyzed™ ACS Reagent