Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Fluoride, Powder, USP, EP, BP, 98.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7681-49-4
FNa
41.99
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride
[F-].[Na+]
7681-49-4
FNa
41.99
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride
[F-].[Na+]

SO167, 7681-49-4, NaF

Spectrum™ Sodium Fluoride, Powder, USP, EP, BP is used to prevent cavities making teeth stronger and more resistant to decay caused by acid and bacteria. Toothpaste being the preferred delivery method. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade Multi Compendia
Acidity 2.0 mL NAOH
CAS Max % 1
Alkalinity 0.5 mL H2SO4
Appearance of Solution Clear
Percent Purity 98.5 to 100.5%
Loss on Drying 0.005
Packaging Poly Pail
Sulfate 200 ppm
Quantity 12 kg