Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Fluoride, Purified, Reagents

Catalog No. 01337020
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 lb.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-020 2.5 kg
01-337-021 125 g
01-337-019 500 g
01-337-018 50 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-020 Supplier Reagents Holdings Llc Supplier No. C23170002.5A5
Only null left
Add to Cart
Add to Cart

Sodium Fluoride, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Boiling Point 1704°C
Melting Point 990°C
Color White
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.78
Chemical Name or Material Sodium Fluoride, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.