Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Formate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

141-53-7
CHNaO2
68.007
HLBBKKJFGFRGMU-UHFFFAOYSA-M
2723810
CHEBI:62965

High quality chemicals for laboratory and specialized industrial use

CAS 141-53-7
Molecular Formula CHNaO2
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES C(=O)[O-].[Na+]
IUPAC Name sodium;formate
Molecular Weight (g/mol) 68.007
PubChem CID 2723810
ChEBI CHEBI:62965
Formula Weight 68.01
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.92kg
Quantity 500g
Chemical Name or Material Sodium Formate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less