Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Gluconate, FCC, 98-102%, Spectrum™

Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
527-07-1
C6H11NaO7
218.14
UPMFZISCCZSDND-UHFFFAOYNA-M
sodium 2,3,4,5,6-pentahydroxyhexanoate
[Na+].OCC(O)C(O)C(O)C(O)C([O-])=O
527-07-1
C6H11NaO7
218.14
UPMFZISCCZSDND-UHFFFAOYNA-M
sodium 2,3,4,5,6-pentahydroxyhexanoate
[Na+].OCC(O)C(O)C(O)C(O)C([O-])=O

SO168, 527-07-1, C6H11NaO7

Spectrum™ Sodium Gluconate, FCC is used as surface cleaning agent for metals and glass bottles. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Quantity 45 kg
CAS Max % 1
Percent Purity 98 to 102%
Grade FCC