Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium glycocholate hydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
207614-05-9
C26H46NNaO8
523.643
MFCD09037360
SDTQNTQLAZRSIC-NWNSWQEHSA-M
glycocholic acid sodium salt hydrate,sodium glycocholate hydrate,c26h42no6.na.2h2o,sodium glycocholate dihydrate
45051920
sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate;dihydrate
CC(CCC(=O)NCC(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.O.O.[Na+]
207614-05-9
C26H46NNaO8
523.643
MFCD09037360
SDTQNTQLAZRSIC-NWNSWQEHSA-M
glycocholic acid sodium salt hydrate,sodium glycocholate hydrate,c26h42no6.na.2h2o,sodium glycocholate dihydrate
45051920
sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate;dihydrate
CC(CCC(=O)NCC(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.O.O.[Na+]
Specific Rotation Condition + 29.50 (20.00°C c=1,H2O)
Infrared Spectrum Authentic
Specific Rotation +28° to +31° (20°C, 589nm) (c=1, H2O) on dry basis
Color Colorless to Yellow
Packaging Glass bottle
Water 8% max. (K.F.)
Quantity 5g
Assay Percent Range 98%
Formula Weight 487.61
Physical Form Liquid
Percent Purity ≥97.5% (on dry basis) (Total base)
Chemical Name or Material Sodium glycocholate hydrate, 98%
Show More Show Less