Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Sodium hexabromoosmate(IV), Premion™, 99.95% (metals basis), Os 26.1% min
Click to view available options
Quantity:
1g
5g
Specifications
Linear Formula | Na2OsBr6 |
Formula Weight | 715.64 |
Physical Form | Crystalline Powder |
Percent Purity | 99.95% |
Grade | Premion™ |
Concentration | Os 26.1% min. |
Color | Black to Red |
Melting Point | (decomposition) |
Quantity | 1g |
Chemical Name or Material | Sodium hexabromoosmate(IV) |
Show More |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures