Alfa Aesar™ Sodium hexabromoplatinate(IV) hexahydrate

Manufacturer: Alfa Aesar™ 8896606

Quantity 5g

 View more versions of this product

Catalog No. AA8896606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 39277-13-9
    Molecular Formula Na2PtBr6·6H2O
    Formula Weight 828.49 (720.49 Anhydrous)
    MDL Number MFCD00014237
    Chemical Name or Material Sodium hexabromoplatinate (IV) hexahydrate

    Specifications

    Formula Weight 828.49 (720.49 Anhydrous)
    Health Hazard 2 GHS H Statement
    H334-H335-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula Na2PtBr6·6H2O
    Physical Form Crystalline
    CAS 39277-13-9
    Quantity 5g
    MDL Number MFCD00014237
    Health Hazard 1 Danger
    Chemical Name or Material Sodium hexabromoplatinate (IV) hexahydrate

    GHS H Statement
    H334-H335-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit