Alfa Aesar™ Sodium hexachloroiridate(IV) hexahydrate, Ir 33.9% min

Manufacturer: Alfa Aesar™ 1189006

Quantity 5g

 View more versions of this product

Catalog No. AA1189006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19567-78-3
    Formula Weight 558.99 (450.92 Anhydrous)
    MDL Number MFCD00149169
    Chemical Name or Material Sodium hexachloroiridate(IV) hexahydrate

    Specifications

    Formula Weight 558.99 (450.92 Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P301+P312-P337+P313-P406-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    If eye irritation persists: Get medical advice/attention.
    Store in corrosive resistant container with a resistant inner liner.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 19567-78-3
    Health Hazard 1 Warning
    Color Red-black
    Melting Point 600°C (dec.)
    Quantity 5g
    Physical Form Powder
    MDL Number MFCD00149169
    Concentration Ir 33.9% min.
    Health Hazard 2 GHS H Statement
    H290-H302-H319
    May be corrosive to metals.
    Harmful if swallowed.
    Causes serious eye irritation.
    Chemical Name or Material Sodium hexachloroiridate(IV) hexahydrate

    GHS H Statement
    H290-H302-H319
    May be corrosive to metals.
    Harmful if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P301+P312-P337+P313-P406-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    If eye irritation persists: Get medical advice/attention.
    Store in corrosive resistant container with a resistant inner liner.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit