Learn More
Sodium hexachloroosmate(IV) dihydrate, Os 38.7% min, Thermo Scientific Chemicals
Description
Sodium hexachloroosmate(IV) dihydrate is used as an intermediate and catalyst in organic synthesis. It is also used to prepare dihydrogen complexes of osmium(II) containing meso-tetraphos-1 ligand.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown to Red |
Physical Form | Crystalline |
Quantity | 5 g |
Linear Formula | Na2OsCl6·2H2O |
Solubility Information | Insoluble in water. |
IUPAC Name | disodium;hexachloroosmium(2-) |
Formula Weight | 484.93 (448.90 Anhydrous) |
Concentration or Composition (by Analyte or Components) | Os 38.7% min. |
Odor | Odorless |
Sensitivity | Air & Moisture Sensitive |
Show More |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 215-152-5 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.