Sodium hexachloroplatinate(IV) hexahydrate, Alfa Aesar™

For Use As a Catalyst

Manufacturer: Alfa Aesar 1104706

Quantity 5g

 View more versions of this product

Catalog No. AA1104706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19583-77-8
    Physical Form Crystalline
    Molecular Formula Na2PtCl6·6H2O
    Formula Weight 561.88 (453.79 Anhydrous)
    MDL Number MFCD00149172
    Synonym Sodium chloroplatinate(IV)
    Chemical Name or Material Sodium hexachloroplatinate(IV) hexahydrate

    Specifications

    Formula Weight 561.88 (453.79 Anhydrous)
    Quantity 5g
    Color Orange
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 240-983-5
    Physical Form Crystalline
    Odor Odorless
    Molecular Formula Na2PtCl6·6H2O
    Appearance Orange powder
    MDL Number MFCD00149172
    CAS 19583-77-8
    Synonym Sodium chloroplatinate(IV)
    Health Hazard 1 H301-H317-H334-H318-H290-H400-H410: Toxic if swallowed. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes serious eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Sodium hexachloroplatinate(IV) hexahydrate

    H301-H317-H334-H318-H290-H400-H410: Toxic if swallowed. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes serious eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit