Alfa Aesar™ Sodium hexachlororhodate(III) dodecahydrate, Rh 17.1%

Manufacturer: Alfa Aesar™ 1180906

Quantity 5g

 View more versions of this product

Catalog No. AA1180906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14972-70-4
    Formula Weight 600.77 (384.59 Anhydrous)
    MDL Number MFCD00003509
    Chemical Name or Material Sodium hexachlororhodate(III) dodecahydrate

    Specifications

    Formula Weight 600.77 (384.59 Anhydrous)
    MDL Number MFCD00003509
    Quantity 5g
    TSCA TSCA
    Physical Form Crystalline
    CAS 14972-70-4
    Melting Point 900°C (dec.)
    Concentration Rh 17.1%
    Chemical Name or Material Sodium hexachlororhodate(III) dodecahydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit