Alfa Aesar™ Sodium hexafluoroaluminate, 97.8+%

Manufacturer: Alfa Aesar™ 8827522

Quantity 100g

 View more versions of this product

Catalog No. AA8827522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13775-53-6
    Molecular Formula Na3AlF6
    Formula Weight 209.95
    MDL Number MFCD00003507
    UN Number UN3260
    Synonym Cryolite; Aluminum sodium fluoride
    Chemical Name or Material Sodium hexafluoroaluminate

    Specifications

    Formula Weight 209.95
    Melting Point 1,000°C
    Health Hazard 3 GHS P Statement
    P260-P261-P304+P340-P301+P312-P312-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Odor Odorless
    UN Number UN3260
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H372-H302-H332
    Causes damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    RTECS Number BD0075000
    Physical Form Powder
    Molecular Formula Na3AlF6
    CAS 13775-53-6
    Density 2.90g/mL
    MDL Number MFCD00003507
    Synonym Cryolite; Aluminum sodium fluoride
    Quantity 100g
    Chemical Name or Material Sodium hexafluoroaluminate
    Assay ≥97.8%

    GHS H Statement
    H372-H302-H332
    Causes damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P304+P340-P301+P312-P312-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit