Alfa Aesar™ Sodium hexafluoroarsenate(V), 99%

Manufacturer: Alfa Aesar™ 8851718

Quantity 50g

 View more versions of this product

Catalog No. AA8851718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12005-86-6
    Molecular Formula NaAsF6
    Formula Weight 211.9
    MDL Number MFCD00014228
    UN Number UN1557
    Chemical Name or Material Sodium hexafluoroarsenate (V)

    Specifications

    Formula Weight 211.9
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 12005-86-6
    UN Number UN1557
    Quantity 50g
    Health Hazard 2 GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.
    Physical Form Powder
    Molecular Formula NaAsF6
    MDL Number MFCD00014228
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
    RTECS Number WB2775000
    Health Hazard 1 Danger
    Chemical Name or Material Sodium hexafluoroarsenate (V)
    Assay 99%

    GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.

    GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit