Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hexafluorophosphate, 98.5+%, pure, Acros Organics

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Plastic bottle
21324-39-0
F6NaP
167.95
MFCD00011122
KMADQUOCJBLXRP-UHFFFAOYSA-N
sodium hexafluorophosphate,sodium hexa-fluoro phosphate,sodium hexafluorophosphate v,napf6,sodium hexaflurophosphate,acmc-1cao2,sodium hexafluoro-,e5-phosphanuide,sodium hexafluoro-$l^ 5-phosphanuide,phosphate 1-, hexafluoro-, sodium 1:1
5147921
sodium;hexafluorophosphate
F[P-](F)(F)(F)(F)F.[Na+]
21324-39-0
F6NaP
167.95
MFCD00011122
KMADQUOCJBLXRP-UHFFFAOYSA-N
sodium hexafluorophosphate,sodium hexa-fluoro phosphate,sodium hexafluorophosphate v,napf6,sodium hexaflurophosphate,acmc-1cao2,sodium hexafluoro-,e5-phosphanuide,sodium hexafluoro-$l^ 5-phosphanuide,phosphate 1-, hexafluoro-, sodium 1:1
5147921
sodium;hexafluorophosphate
F[P-](F)(F)(F)(F)F.[Na+]
C-X bond formation reagent
Assay Percent Range 98.5% min
Linear Formula NaPF6
Solubility Information Solubility in water: soluble
Formula Weight 167.95
Physical Form Crystalline Powder
Percent Purity 98.5+%
Grade Pure
Packaging Plastic bottle
Solubility Solubility in water: soluble.
Color White to Gray
Melting Point 200.0°C
Quantity 250g
Chemical Name or Material Sodium hexafluorophosphate
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

EINECSNumber : 244-333-1