Sodium hexafluorophosphate, >99%, Alfa Aesar™

Manufacturer: Alfa Aesar 1306414

Quantity 25g

 View more versions of this product

Catalog No. AA1306414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21324-39-0
    Physical Form Crystals
    Assay Percent Range >99%
    Molecular Formula NaPF6
    Formula Weight 167.95
    MDL Number MFCD00011122
    Chemical Name or Material Sodium hexafluorophosphate

    Specifications

    Physical Form Crystals
    Formula Weight 167.95
    Quantity 25g
    Color White
    DOT Information Hazard Class: 8, Packing Group: II
    EINECS Number 244-333-1
    Assay Percent Range >99%
    Molecular Formula NaPF6
    MDL Number MFCD00011122
    CAS 21324-39-0
    Odor Odorless
    Appearance White crystalline
    Health Hazard 1 H314-H318-H302-H312-H332: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.
    Chemical Name or Material Sodium hexafluorophosphate

    H314-H318-H302-H312-H332: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit