Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium hexanitritoiridate(III), 99%

Click to view available options
Quantity:
0.1g
0.5g
2.5g
94022-51-2
IrN6Na3O12
537.22
MFCD00064621
GKIZAPPVQAUTGW-UHFFFAOYSA-H
Sodium hexakis(nitrito)iridate(III)
94022-51-2
IrN6Na3O12
537.22
MFCD00064621
GKIZAPPVQAUTGW-UHFFFAOYSA-H
Sodium hexakis(nitrito)iridate(III)
Linear Formula Na3Ir(NO2)6
Formula Weight 537.22
Percent Purity 99%
Quantity 0.5g
Chemical Name or Material Sodium hexanitritoiridate(III)

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning