Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium hydride, ca 60% dispersion in oil, in solvent solublebags

Manufacturer:  Alfa Aesar™ H3649022

Quantity 100g

 View more versions of this product

Catalog No. AAH3649022


  Check Availability
Add to cart

Specifications

Description

Applications
Sodium hydride can be used for direct intercalation into C60, resulting in superconducting material (NaH)4C60. It is primarily used as a strong base in organic synthesis. It can be used to dry some organic solvents. It has been proposed for hydrogen storage for use in fuel cell vehicles.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, acid chlorides, acids, halogens and alcohols.
Description & Specifications

Chemical Identifiers

7646-69-7
MFCD00003471
Sodium hydride, ≈60% dispersion in oil, in solvent solublebags
24
UN1427

Specifications

7646-69-7
24
UN1427
Air & Moisture Sensitive
100g
Moist Powder
MFCD00003471
Odorless
White to pale gray
Sodium hydride, ≈60% dispersion in oil, in solvent solublebags

Safety and Handling

SDS & Safety Documents

GHS H Statement
H260
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: SODIUM HYDRIDE

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit