Alfa Aesar™ Sodium hydride, ca 60% dispersion in oil, in solvent solublebags

Manufacturer: Alfa Aesar™ H3649036

Quantity 500g

 View more versions of this product

Catalog No. AAH3649036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7646-69-7
    Formula Weight 24
    MDL Number MFCD00003471
    UN Number UN1427
    Chemical Name or Material Sodium hydride, ≈60% dispersion in oil, in solvent solublebags

    Specifications

    Formula Weight 24
    Health Hazard 3 GHS P Statement
    P231+P232-P280-P233-P370+P378c-P402+P404-P501a
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep container tightly closed.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Moist Powder
    Sensitivity Air & Moisture Sensitive
    MDL Number MFCD00003471
    UN Number UN1427
    Color White to pale gray
    Quantity 500g
    Health Hazard 1 Danger
    CAS 7646-69-7
    Health Hazard 2 GHS H Statement
    H260
    In contact with water releases flammable gases which may ignite spontaneously.
    DOT Information Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: SODIUM HYDRIDE
    Odor Odorless
    Chemical Name or Material Sodium hydride, ≈60% dispersion in oil, in solvent solublebags

    Description

    Applications
    Sodium hydride can be used for direct intercalation into C60, resulting in superconducting material (NaH)4C60. It is primarily used as a strong base in organic synthesis. It can be used to dry some organic solvents. It has been proposed for hydrogen storage for use in fuel cell vehicles.

    Solubility
    It reacts with water.

    Notes
    Air and moisture sensitive. Incompatible with oxidizing agents, acid chlorides, acids, halogens and alcohols.

    GHS H Statement
    H260
    In contact with water releases flammable gases which may ignite spontaneously.

    GHS P Statement
    P231+P232-P280-P233-P370+P378c-P402+P404-P501a
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep container tightly closed.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit