Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydrogen arsenate heptahydrate, ACS, 98.0 to 102.0%, Alfa Aesar™

Manufacturer:  Alfa Aesar 3337336

Quantity 500g

 View more versions of this product

Catalog No. AA3337336


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

10048-95-0
98.0-102.0%
312.01; 185.94 (anhydrous)
Arsenic acid disodium salt
ACS Grade
Granular
HAsNa2O4·7H2O
MFCD00149179
Sodium hydrogen arsenate heptahydrate, For analysis ACS

Specifications

10048-95-0
98.0-102.0%
White granular
MFCD00149179
White
Sodium hydrogen arsenate heptahydrate, For analysis ACS
Granular
HAsNa2O4·7H2O
312.01; 185.94 (anhydrous)
Arsenic acid disodium salt
500g
ACS Grade

Safety and Handling

SDS & Safety Documents

H301-H331-H350-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause cancer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1; Packing Group: II

EINECSNumber : 231-902-4

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit