Learn More
Sodium hydrogen arsenate heptahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Description
Sodium hydrogen arsenate heptahydrate is used as a source of soluble arsenic and in dyeing and printing with Turkey-red oil. It also acts as a veterinary drug for anti-parasitic, skin diseases, blood diseases, rheumatism and asthma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 130°C (decomposition) |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 25 g |
Assay Percent Range | 98.0-102.0% |
Solubility Information | Soluble in water and glycerol. |
IUPAC Name | disodium heptahydrate hydrogen arsorate |
Formula Weight | 312.01 (185.94 Anhydrous) |
Grade | ACS |
Appearance | White granular |
Show More |
Safety and Handling
Hazard Category | H301+H331-H350 |
Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c |
DOTInformation | DOT Class: 6.1; Packing Group: II |
EINECSNumber | 231-902-4 |
RTECSNumber | CG0900000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.