Learn More
Sodium hydrogen carbonate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Sodium hydrogen carbonate is used in a process for removing paint and corrosion called sodablasting; the process is particularly suitable for cleaning aluminum panels which can be distorted by other types of abrasive. It can be administered to pools, spas, and garden ponds to raise pH levels. Also can be used as a fire extinguisher. Its primary use is in cooking (baking), as a leavening agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 99.998% (metals basis) |
Beilstein | 4153970 |
Solubility Information | Soluble in water. Insoluble in ethanol. |
IUPAC Name | sodium hydrogen carbonate |
Formula Weight | 84.01 |
Odor | Odorless |
Refractive Index | 1.5 |
Show More |
Safety and Handling
Hazard Category | H303: May be harmful if swallowed. |
EINECSNumber | 205-633-8 |
RTECSNumber | VZ0950000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.